Learn More
ENG Scientific Manual Method Stain Buffer Solution
Description
- This solution, using a flooding technique, will stain all cellular elements without precipitate or artifactural changes.
- Staining characteristics are similar to H-PACK.
- Buffer is applied immediately after stain, reducing overall staining time.
- Convenient flip-top cap for 250 mL and 950 mL sizes.
- IN THE EVENT OF COLD WEATHER, allow solution to return to room temperature and SHAKE WELL BEFORE USE.
- Contents: Wright Stain, Giemsa Stain, Methanol, Buffers, Stabilizers.
- These solutions are made from certified dyes (when applicable).
Specifications
Specifications
| Quantity | 950 mL |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For examining Bloody specimen material or Blood Culture supernatant fluid, in-vitro diagnostic use |
| pH | 6.73 to 6.77 |
| Packaging | Bottle |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.