Learn More
ENG Scientific Manual Method Wright Giemsa Stain
Description
- Manual Method Stain (Wright or Wright Giemsa)
- Available with and without matching buffer solution
- Buffer is applied immediately after stain, reducing overall staining time
- Convenient flip top cap for 250 mL and 950 mL sizes
- Solutions made from certified dyes
- Staining time: 5 to 7 minutes, depending on color intensity desired
- During cold weather, allow solutions to return to room temperature and shake well before use
Specifications
Specifications
| Quantity | 2 x 950 mL |
| For Use With (Application) | In vitro diagnostic use |
| Staining Time | 5 to 7 min. |
| Packaging | Bottles |
| Product Type | Wright Giemsa Stain |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.