Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Mayer's Mucicarmine Stain

Catalog No. ES9161
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9161 100 mL
ES9162 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES9161 Supplier ENG Scientific Supplier No. 9161
Add to Cart
Add to Cart

Mayer's Mucicarmine Stain is designed to show mucin derived from epithelial cells (Bronchi and Colon).

  • Includes Carmine, Aluminum Chloride
  • Solution made from certified dyes

Specifications

Content And Storage Carmine, Aluminum Chloride
Packaging Type Bottle
Product Type Staining Reagent
Quantity 100 mL
For Use With (Application) In vitro diagnostic use only
Test Time 15 min.

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.