Learn More
ENG Scientific Methanol Fixative
Description
- Methanol preserves the morphology of red blood cells as well as bacteria
Specifications
Specifications
| Quantity | 2 x 950 mL |
| For Use With (Application) | For use as a fixative prior to staining bloody specimen material or blood culture supernatant fluid |
| Stain Type | Methanol |
| Product Type | Methanol Fixative |
| Packaging | Bottle |
For in vitro diagnostic use only.