Learn More
ENG Scientific Methyl Green with Pyronin Stain
Catalog No. ES9411
Change view
Click to view available options
Quantity:
1 x 250 mL
2 x 250 mL
2 x 950 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9411 2 x 250 mL
ES9410 1 x 250 mL
ES9412 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES9411 Supplier ENG Scientific Supplier No. 9411
Only null left
Add to Cart
Add to Cart
Methyl Green with Pyronin Stain uses the Modified Unna Pappenheim Method by Saathof for bacteria, DNA, RNA and Plasma cells. Alcohol-ether fixed.
  • Smears are brought to distilled water through alcohols.
Specifications
Content And Storage Methyl Green, Pyronin, Ethanol, Glycerol, Phenol
Packaging Type Pack (Bottles)
Product Type Staining Reagent
Quantity 2 x 250 mL
For in vitro diagnostic use only.