Learn More
Eng Scientific Methylene Blue Solution
Catalog No. ES4870
Change view
Click to view available options
Quantity:
100 mL
3.8 L
Packaging:
1 Bottle
1 Pack (2 Bottles)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ES4870 100 mL 1 Pack (2 Bottles)
ES4871 100 mL 1 Bottle
ES4879 3.8 L 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES4870 Supplier Eng Scientific Inc Supplier No. 4870
Add to Cart
Add to Cart
Solutions; Methylene Blue; Eng Scientific; Counterstain or wet mount; For immediate examination of fecal sample for trophozoites; 3 percent aqueous; 3.8L
Specifications
Packaging 1 Pack (2 Bottles)
Quantity 100 mL