Learn More
ENG Scientific Phloxine B Counterstain
Description
- Solution is used mainly as background contrast to nuclear stains, i.e., hematoxylin-eosin sequence.
- Available in 1% and 0.05% solution.
- Made from certified dyes.
- For in vitro diagnostic use only.
Specifications
Specifications
| For Use With (Application) | Solution is used mainly as background contrast to nuclear stains, i.e., hematoxylin eosin sequence |
| Includes | Phloxine, Calcium Chloride |
| Quantity | 1 x 950 mL (32 oz.) |
| Product Type | Phloxine B Counterstain (1%) |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.