Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Platelet Diluent

Catalog No. ES5300
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES5300 2 x 100 mL
ES5301 1 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES5300 Supplier ENG Scientific Supplier No. 5300
Add to Cart
Add to Cart

For platelet counts.

  • Contains Ammonium Oxalate
  • For in vitro use only

Specifications

Quantity 2 x 100 mL
For Use With (Application) For platelet counts, in-vitro diagnostic use only
Staining Time 15 to 20 min.
Packaging Bottles

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.