Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Pneumocystis Carinii Stain Kit

Catalog No. ES7152
Change view
Click to view available options
Quantity:
2 x 250 mL
4 x 250 mL
Product Type:
Ethanol 95%
Modified Toluidine Blue O Stain
Xylene
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES7152 2 x 250 mL Ethanol 95%
ES7150 4 x 250 mL Modified Toluidine Blue O Stain
ES7154 2 x 250 mL Xylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES7152 Supplier ENG Scientific Supplier No. 7152
Add to Cart
Add to Cart

Modified Toluidine Blue O Stain allows the Pneumocystis carinii cysts to be visualized more easily after staining. For Pneumocystis Carinii Stain Kit (Modified Toluidine Blue O). Includes: 250 mL each of Solutions I, II, III and IV.

  • Pneumocystis carinii is an opportunist which causes a diffuse interstitial pneumonia in patients with impaired immune systems.
  • The disease itself can be classified as epidemic or sporadic; the latter occurring in patients who have an underlying immunosuppression.
  • In this procedure, a sulfation reagent of glacial acetic acid and sulfuric acid (not included) is used for the removal of background material.
  • This allows the Pneumocystis carinii cysts to be visualized more easily after staining with Toluidine Blue 0.
  • Diagnosis of Pneumocystis carinii pneumonia can frequently be made on bronchoalveolar lavage (BAL) or on touch preparations of pulmonary tissue (open lung biopsies and transbronchial biopsies).
  • Recommended Procedures:
    • BAL Specimens:
      • Using a 50 mL tube, centrifuge for 15 minutes at 2000 x g.
      • Aspirate all but the bottom 5 mL of supernatant.
      • With a Pasteur pipette, aspirate the sediment plus approximately 1 mL of the remaining 5 mL of fluid.
      • Transfer material to a 15 mL centrifuge tube, gently mix and prepare smear by spreading a drop over a 1 cmm area.
      • If concentrated specimen is very thick or mucoid, spread over entire slide with care being taken not to make the smear too thick.
      • Dry the slides on a heating block at 50 - 55°C.
      • Allow to cool before staining.
    • Touch Preparations:
      • When dry, process in same manner as slides of BAL.
      • Preparation of Sulfation Reagent:
        • In a dry coplin jar submerged in a plastic tub filled with cool water (not below 10°C), mix 45 mL of glacial acetic acid with 15 mL of concentrated sulfuric acid and stir gently with glass rod.
      • Carefully place smears in reagent for 10 minutes.
      • Mix immediately and again after 5 minutes.
      • Gently rinse with water for 10 minutes.
      • Drain excess water and stain with Sol. I for 3 - 5 minutes.
      • Rinse with Sol. II followed by Sol. III for 10 seconds each.
      • Clear in Sol. IV, two changes each for 10 seconds and mount.
  • Results: The cyst forms appear as lavender structures (cup-shaped) approximately 5 urn in diameter. The cyst outline is distinct, and the internal region stains uniformly.
  • Note: A negative bronchoalveolar lavage does not rule out an infection with Pneumocystis carinii. Other diagnostic procedures may be necessary.

Specifications

Quantity 2 x 250 mL
Includes Toluidine Blue O Stain
For Use With (Application) In vitro diagnostic use
Product Type Ethanol 95%

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.