Learn More
ENG Scientific Potassium Hydroxide 10%
Description
- Potassium Hydroxide (10%)
Specifications
Specifications
| Type | Potassium Hydroxide (10%) |
| Packaging Type | Bottle |
| Quantity | 1 x 475 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.