Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific RBC Diluting Fluid (Gower's Solution)

Catalog No. ES5602
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES5602 2 x 100 mL
ES5603 1 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES5602 Supplier ENG Scientific Supplier No. 5602
Add to Cart
Add to Cart

RBC Diluting Fluid (Gower's Solution) is used for diluting RBC's. Contains Sodium sulfate and Glacial acetic acid. Settling time is between 1 to 3 minutes.

  • Contains: Sodium sulfate, Glacial acetic acid

Specifications

Quantity 2 x 100 mL
For Use With (Application) For in-vitro diagnostic use only
Staining Time 1 to 3 min.
Packaging Bottles

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.