Learn More
ENG Scientific Reticulocyte Stain
Description
- The proportion of blood to stain should be increased for low hemoglobins.
- May be used for finger sticks or anticoagulated blood.
Specifications
Specifications
| Quantity | 1 x 100 mL |
| For Use With (Application) | For in-vitro diagnostic use only, may be used for finger sticks or anticoagulated Blood |
| Packaging | Bottle |
| Product Type | BCB |
For in vitro diagnostic use only.