Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Siderocyte Stain Kit, Potassium Ferrocyanide (2%)

Catalog No. ES3481
Change view
Click to view available options
Quantity:
1 x 250 mL
2 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES3481 1 x 250 mL
ES3484 2 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES3481 Supplier ENG Scientific Supplier No. 3481
Add to Cart
Add to Cart

Used in Hematology. Siderocyte Stain Kit for Prussian Blue reaction. Siderotic granules stain bright blue.

  • Mixed with solution II to obtain Prussian blue reaction

Specifications

Quantity 1 x 250 mL
For Use With (Application) For in-vitro diagnostic use only
Packaging Bottle

For in vitro diagnostic use only. Due to product restrictions, please Sign In to purchase or view availability for this product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.