Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solution I, Carbol Fuchsin

Catalog No. ES6810
Change view
Click to view available options
Quantity:
1 x 250 mL
1 x 3800 mL (1 gal.)
2 x 250 mL
2 x 950 mL
For Use With (Application):
Acid-fast stain kit
Cryptosporidium stain kit
Cyclospora stain kit
Spore stain kit
Ziehl-Neelsen Stain Kit
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity For Use With (Application)
ES6810 1 x 250 mL Spore stain kit
ES6466 2 x 250 mL Cyclospora stain kit
ES6471 2 x 250 mL Cryptosporidium stain kit
ES6531 2 x 250 mL Acid-fast stain kit
ES7010 2 x 250 mL Ziehl-Neelsen Stain Kit
ES7020 2 x 950 mL Ziehl-Neelsen Stain Kit
ES7021 1 x 3800 mL (1 gal.) Ziehl-Neelsen Stain Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. ES6810 Supplier ENG Scientific Supplier No. 6810
Add to Cart
Add to Cart

Solution I, Carbol Fuchsin contains Basic Fuchsin, Ethanol, Phenol, Sulfuric Acid, Malachite Green. For use with Cyclospora Stain kit.

  • Solution I - Carbol Fuchsin
  • Solution II - Acid Alcohol (10% Sulfuric)
  • Solution III - Malachite Green Contents: Basic Fuchsin, Ethanol, Phenol, Sulfuric Acid, Malachite Green
  • Solutions made from certified dyes (when applicable)

Specifications

Quantity 1 x 250 mL
For Use With (Application) Spore stain kit
Composition Basic Fuchsin, Phenol, Ethanol, Acetic Acid, Methylene Blue, Malachite Green
Product Type Carbol Fuchsin

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.