Learn More
ENG Scientific Solution II - Acid Alcohol
Description
- Solution II - Acid Alcohol
Specifications
Specifications
| For Use With (Application) | Ideal for staining acid-fast bacteria |
| For Use With (Equipment) | Acid-fast Stain Kit (Modified Kinyoun Method) |
| Packaging | Bottle |
| Quantity | 2 x 950 mL |
| Product Type | Solution II - Acid Alcohol |
For in vitro diagnostic use only.