Learn More
ENG Scientific Solution III - Loeffler's Methylene Blue
Description
- Solution III - Loeffler's Methylene Blue
Specifications
Specifications
| For Use With (Application) | Ideal for staining of acid-fast bacteria |
| For Use With (Equipment) | Ziehl-Neelsen Stain Kit |
| Packaging | Bottles |
| Quantity | 2 x 950 mL |
| Product Type | Solution III - Loefflers Methylene Blue |
For in vitro diagnostic use only.