Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solution III - Modified Trichrome Stain

Catalog No. ES6650
Change view
Click to view available options
Quantity:
1 x 250 mL
1 x 950 mL
2 x 250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6650 1 x 250 mL
ES6651 2 x 250 mL
ES6652 1 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES6650 Supplier ENG Scientific Supplier No. 6650
Add to Cart
Add to Cart

For quick detection of Microsporidia Spores in Stool and Duodenal Aspirates. Non-evasive method for Modified Trichrome Stain Kit.

  • Solution I - Formalin (10%)
  • Solution II - Methanol Fixative
  • Solution III - Modified Trichrome Stain
  • Solution IV - Acid Alcohol
  • Solution V - Ethanol (95%)
  • Solution VI - Ethanol
  • Solution VII - Xylene
  • This stain method was designed as a simple, non-invasive technique for the rapid detection of microsporidia spores in fecal material by light microscopy.
  • The chromotrope based stain is a modification of the trichrome stain for intestinal protozoa.
  • Contents: Chromotrope 2R, Fast Green FCF, Phosphotungstic Acid, Glacial Acetic Acid.
  • These solutions are made from certified dyes (when applicable).

Specifications

For Use With (Application) Ideal for Microsporidia spores in stool and duodenal aspirates
Packaging Bottle
Quantity 1 x 250 mL
Product Type Solution III - Modified Trichrome Stain

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.