Learn More
ENG Scientific Solution IIIA - Brilliant Green
Description
- Use with Acid-fast stain kit.
- Modification of the Kinyoun Method.
Specifications
Specifications
| For Use With (Application) | Acid-fast stain kit |
| Quantity | 2 x 250 mL |
| Product Type | Solution IIIA - Brilliant Green |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.