Learn More
ENG Scientific Solution IIIA - Ethanol (95%)
Description
- Available in various counterstains and decolorizers for Gram Stain Kit (For Differential Staining of Bacteria)
- Kit contains Solution IIIA - 95% Ethanol
Specifications
Specifications
| Concentration | 95% |
| For Use With (Application) | Ideal for differential staining of bacteria |
| Packaging | Bottles |
| Quantity | 4 x 250 mL |
| Product Type | Solution IIIA - Ethanol |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.