Learn More
ENG Scientific Stat-Quick Wright and Wright Giemsa Stain Kits
Catalog No. ES4380
Change view
Click to view available options
Quantity:
2 x 3800 mL (1 gal.)
2 x 950 mL
Product Type:
Wright Giemsa Stain and Matching Buffer
Wright Stain and Matching Buffer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES4380 2 x 950 mL Wright Giemsa Stain and Matching Buffer
ES4350 2 x 950 mL Wright Stain and Matching Buffer
ES4360 2 x 3800 mL (1 gal.) Wright Stain and Matching Buffer
ES4390 2 x 3800 mL (1 gal.) Wright Giemsa Stain and Matching Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ES4380 Supplier ENG Scientific Supplier No. 4380
Add to Cart
Add to Cart
Utilized in both stat and routine laboratories. For hematology staining. Stat Quick Stain (Wright or Wright Giemsa) and Buffers.
  • Only two solutions needed to obtain excellent staining
  • No fixation required with this stain
  • With matching buffer solution
  • Matching buffer solution allows uniform staining of all cellular elements without precipitate or artifactual changes
  • Stain placed in staining container should be mixed periodically
  • During cold weather, allow solution to return to room temperature and shake well before use
  • Used in both stat and routine laboratories
Specifications
Quantity 2 x 950 mL
Packaging Kit
Product Type Wright Giemsa Stain and Matching Buffer
For in vitro diagnostic use only.