Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Stat-Quick Wright and Wright Giemsa Stains and Buffers

Catalog No. ES4320
Change view
Click to view available options
Quantity:
1 x 19 L (5 gal.)
1 x 3.8 L (1 gal.)
1 x 950 mL
2 x 950 mL
Product Type:
Buffer
Wright Giemsa Stain
Wright Stain
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES4320 2 x 950 mL Wright Giemsa Stain
ES4301 1 x 950 mL Wright Stain
ES4300 2 x 950 mL Wright Stain
ES4310 1 x 3.8 L (1 gal.) Wright Stain
ES4370 1 x 3.8 L (1 gal.) Buffer
ES4330 1 x 3.8 L (1 gal.) Wright Giemsa Stain
ES4371 1 x 19 L (5 gal.) Buffer
ES4321 1 x 950 mL Wright Giemsa Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. ES4320 Supplier ENG Scientific Supplier No. 4320
Add to Cart
Add to Cart
This item is not returnable. View return policy

Stat-Quick Wright and Wright Giemsa Stains and Buffers are utilized in both stat and routine laboratories. For hematology staining. Stat Quick Stain (Wright or Wright Giemsa) and Buffers.

  • No fixation required with stains
  • Matching buffer solution allows uniform staining of all cellular elements without precipitate or artifactual changes
  • During cold weather, allow solution to return to room temperature and shake well before use

Specifications

Quantity 2 x 950 mL
For Use With (Application) Hematology Staining
Packaging Bottles
Product Type Wright Giemsa Stain

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.