Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Sudan Black B Stain Kit

Catalog No. ES4470
Change view
Click to view available options
Quantity:
2 x 250 mL
2 x 950 mL
4 x 250 mL
Product Type:
Solution I Sudan Black B
Solution II Buffer
Solution III Alcohol (Ethanol, 70%)
Solution IV Harris Alum Hematoxylin
Sudan Black B Stain Kit
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES4470 2 x 250 mL Solution IV Harris Alum Hematoxylin
ES4420 2 x 950 mL Solution I Sudan Black B
ES4400 4 x 250 mL Sudan Black B Stain Kit
ES4410 2 x 250 mL Solution I Sudan Black B
ES4450 2 x 250 mL Solution III Alcohol (Ethanol, 70%)
ES4440 2 x 950 mL Solution II Buffer
ES4430 2 x 250 mL Solution II Buffer
ES4460 2 x 950 mL Solution III Alcohol (Ethanol, 70%)
ES4480 2 x 950 mL Solution IV Harris Alum Hematoxylin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. ES4470 Supplier ENG Scientific Supplier No. 4470
Add to Cart
Add to Cart
This item is not returnable. View return policy

For differential diagnosis of acute granulocytic leukemia, acute lymphocytic and acute myelo-monocytic leukemia. For Sudan Black B Stain.

  • Contains Sudan Black B, ethanol, buffers, stabilizers, hematoxylin, potassium alum, mercuric oxide

Specifications

For Use With (Application) For differential diagnosis of acute granulocytic leukemia, acute lymphocytic and acute myelo-monocytic leukemia
Packaging Bottles
Quantity 2 x 250 mL
Product Type Solution IV Harris Alum Hematoxylin

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.