Learn More
ENG Scientific Unna Stain
Description
- Includes Methylene Blue, Ethanol, Potassium Carbonate
- Made from certified dyes
- For in vitro diagnostic use only
Specifications
Specifications
| For Use With (Application) | Used in histology for the differentiation of mast cells from plasma cells |
| Includes | Methylene Blue, Ethanol, Potassium Carbonate |
| Packaging | Bottles |
| Quantity | 2 x 100 mL |
| Product Type | Unna Stain |
For in vitro diagnostic use only.