Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Urinary Sediment Stain

Catalog No. ES3151
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES3151 1 x 100 mL
ES3150 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES3151 Supplier ENG Scientific Supplier No. 3151

A quick staining procedure by which a drop of stain solution is directly added to urine sediment.

  • Modification of Sternheimer and Malbin
  • Contents: Phloxine, Propylene Glycol, Sodium Carbonate Phloxine, Propylene Glycol, Sodium Carbonate
  • Solution made from certified dyes

Specifications

Type Urinary Sediment Stain
For Use With (Application) Visualize neutrophils, glitter and squamous cells, bacteria and hyaline, granular and cellular casts
Includes Crystal violet, ammonium oxalate, ethanol, safranin
Packaging Type Bottle
Quantity 1 x 100 mL

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.