Learn More
ENG Scientific Verhoeff's Elastic Tissue Stain Kit
Description
Verhoeff's Elastic Tissue Stain Kit is used to stain elastic fibers, nuclei and collagen.
- Stains elastic fibers - intensely blue-black, nuclei - blue to black, collagen - red
- Made from certified dyes (when applicable)
- Kit contents: Solution I - Alcoholic Hematoxylin, Solution II - Aqueous Ferric Chloride (10%), Solution III - Iodine, Solution IV - Aqueous Ferric Chloride (2%), and Solution V - Aqueous Phloxine (0.5%)
- Verhoeff's Elastic Tissue Stain Kit
- Solutions also available separately
- For in vitro diagnostic use only
- Used to stain elastic fiber, nuclei and collagen
Specifications
Specifications
| For Use With (Application) | Used to stain elastic fiber, nuclei and collagen |
| Includes | Solution I - Alcoholic Hematoxylin, Solution II - Aqueous Ferric Chloride (10%), Solution III - Iodine, Solution IV - Aqueous Ferric Chloride (2%), and Solution V - Aqueous Phloxine (0.5%) |
| Packaging | Complete Kit (Bottles) |
| Quantity | 5 x 950 mL |
| Product Type | Verhoeff's Elastic Tissue Stain Kit |
For in vitro diagnostic use only.