Learn More
ENG Scientific Verhoeff's Elastic Tissue Stain Kit
Catalog No. ES9280
Change view
Click to view available options
Product Type:
Solution I Alcoholic Hematoxylin
Solution II Aqueous Ferric Chloride
Solution III Iodine
Solution IV Aqueous Ferric Chloride
Solution V Aqueous Phloxine
Verhoeff's Elastic Tissue Stain Kit
Quantity:
2 x 950 mL
5 x 250 mL
5 x 950 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
ES9280 Solution II Aqueous Ferric Chloride 2 x 950 mL
ES9260 Verhoeff's Elastic Tissue Stain Kit 5 x 950 mL
ES9250 Verhoeff's Elastic Tissue Stain Kit 5 x 250 mL
ES9290 Solution III Iodine 2 x 950 mL
ES9300 Solution IV Aqueous Ferric Chloride 2 x 950 mL
ES9270 Solution I Alcoholic Hematoxylin 2 x 950 mL
ES9310 Solution V Aqueous Phloxine 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
Catalog No. ES9280 missing translation for 'mfr' ENG Scientific missing translation for 'supplierNo' 9280
Add to Cart
Add to Cart
Verhoeff's Elastic Tissue Stain Kit is used to stain elastic fibers, nuclei and collagen.
  • Stains elastic fibers - intensely blue-black, nuclei - blue to black, collagen - red
  • Made from certified dyes (when applicable)
  • Kit contents: Solution I - Alcoholic Hematoxylin, Solution II - Aqueous Ferric Chloride (10%), Solution III - Iodine, Solution IV - Aqueous Ferric Chloride (2%), and Solution V - Aqueous Phloxine (0.5%)
  • Verhoeff's Elastic Tissue Stain Kit
  • Solutions also available separately
  • For in vitro diagnostic use only
  • Used to stain elastic fiber, nuclei and collagen
Specifications
For Use With (Application) Used to stain elastic fiber, nuclei and collagen
Packaging Pack (Bottles)
Quantity 2 x 950 mL
Product Type Solution II Aqueous Ferric Chloride
For in vitro diagnostic use only.