Learn More
ENG Scientific Verhoeff's Elastic Tissue Stain Kit
Catalog No. ES9300
Change view
Click to view available options
Product Type:
Solution I Alcoholic Hematoxylin
Solution II Aqueous Ferric Chloride
Solution III Iodine
Solution IV Aqueous Ferric Chloride
Solution V Aqueous Phloxine
Verhoeff's Elastic Tissue Stain Kit
Quantity:
2 x 950 mL
5 x 250 mL
5 x 950 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
ES9300 Solution IV Aqueous Ferric Chloride 2 x 950 mL
ES9260 Verhoeff's Elastic Tissue Stain Kit 5 x 950 mL
ES9250 Verhoeff's Elastic Tissue Stain Kit 5 x 250 mL
ES9290 Solution III Iodine 2 x 950 mL
ES9280 Solution II Aqueous Ferric Chloride 2 x 950 mL
ES9270 Solution I Alcoholic Hematoxylin 2 x 950 mL
ES9310 Solution V Aqueous Phloxine 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. ES9300 Supplier ENG Scientific Supplier No. 9300
Add to Cart
Add to Cart
Verhoeff's Elastic Tissue Stain Kit is used to stain elastic fibers, nuclei and collagen.
  • Stains elastic fibers - intensely blue-black, nuclei - blue to black, collagen - red
  • Made from certified dyes (when applicable)
  • Kit contents: Solution I - Alcoholic Hematoxylin, Solution II - Aqueous Ferric Chloride (10%), Solution III - Iodine, Solution IV - Aqueous Ferric Chloride (2%), and Solution V - Aqueous Phloxine (0.5%)
  • Verhoeff's Elastic Tissue Stain Kit
  • Solutions also available separately
  • For in vitro diagnostic use only
  • Used to stain elastic fiber, nuclei and collagen
Specifications
For Use With (Application) Used to stain elastic fiber, nuclei and collagen
Packaging Pack (Bottles)
Quantity 2 x 950 mL
Product Type Solution IV Aqueous Ferric Chloride
For in vitro diagnostic use only.