Learn More
ENG Scientific Weigert's Iron Hematoxylin Stain Kit
Catalog No. ES8600
Change view
Click to view available options
Quantity:
1 x 950 mL
2 x 250 mL
2 x 3800 mL (1 gal.)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8600 2 x 250 mL
ES8602 2 x 3800 mL (1 gal.)
ES8601 1 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. ES8600 Supplier ENG Scientific Supplier No. 8600
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes Solution I - Weigert's Hematoxylin and Solution II - Weigert's Iron Chloride. Kit is available in three sizes.
  • Solution I is Weigert's Hematoxylin
  • Solution II is Weigert's Iron Chloride
  • To create working solution, mix equal parts of Solutions I and II
  • Recommended staining time is 2 - 5 minutes
  • Nuclei stains black, connective tissue stains red, and cytoplasm stains yellowish
Specifications
Includes Bottle of each solution (Hematoxylin, Ethanol, Ferric Chloride, Hydrochloric Acid)
Packaging Complete Kit (Bottles)
Quantity 2 x 250 mL
Product Type Weigert's Iron Hematoxylin Stain Kit