Learn More
ENG Scientific Weigert's Iron Hematoxylin Stain Kit
Catalog No. ES8601
Change view
Click to view available options
Quantity:
1 x 950 mL
2 x 250 mL
2 x 3800 mL (1 gal.)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8601 1 x 950 mL
ES8600 2 x 250 mL
ES8602 2 x 3800 mL (1 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES8601 Supplier ENG Scientific Supplier No. 8601
Add to Cart
Add to Cart
Includes Solution I - Weigert's Hematoxylin and Solution II - Weigert's Iron Chloride. Kit is available in three sizes.
  • Solution I is Weigert's Hematoxylin
  • Solution II is Weigert's Iron Chloride
  • To create working solution, mix equal parts of Solutions I and II
  • Recommended staining time is 2 - 5 minutes
  • Nuclei stains black, connective tissue stains red, and cytoplasm stains yellowish
Specifications
Includes Bottle of each solution (Hematoxylin, Ethanol, Ferric Chloride, Hydrochloric Acid)
Packaging Complete Kit (Bottles)
Quantity 1 x 950 mL
Product Type Weigert's Iron Hematoxylin Stain Kit