Learn More
ENG Scientific Wright Giemsa Quick Stain Kit
Catalog No. ES4100
Change view
Click to view available options
Quantity:
3 x 3800 mL (1 gal.)
3 x 475 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES4100 3 x 475 mL
ES4110 3 x 3800 mL (1 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES4100 Supplier ENG Scientific Supplier No. 4100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
For excellent staining results in one minute without precipitate. Alcoholic quick stain technique. No fixation necessary. For Quick Stain Kit (Wright or Wright Giemsa).
  • Less time in Solution III provides a more alkaline (blue) stain
  • Increase time for a more acidic (red) stain
  • For excellent staining results in one minute
Specifications
Quantity 3 x 475 mL
For Use With (Application) For excellent staining results in one min.
Packaging Complete Kit (Bottles)
For in vitro diagnostic use only.