Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Engino™ STEM Fluid Dynamics
SDPSCI_ED

Catalog No. S000839 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000839 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking Hazard
Catalog No. S000839 Supplier United Scientific™ Supplier No. STEM45
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking Hazard

Learn all about fluid dyanmics with the Engino STEM Fluid Dyanmics Kit.

One of the most important areas in physics is the study of fluids and their fundamental principals. Water and air are the two most important elements for life and both are categorized as fluids. Air motion is what determines the temperature in our rooms. Pipelines are providing water to our homes. Industrial and construction machines use hydraulics to easily lift heavy objects. Airplanes and ships change direction by using aerodynamics and hydrodynamics respectively. Have you ever wondered why objects sink in water, while others float? How does blood flow in our body? How do water tanks operate? With this set you can build 6 different models, including a boat and a submarine, as well as various experimental rigs to learn all about fluid dynamics! Printed building instructions for 4 models are included and 2 more models are available online and through the free kid CAD app for smart devices. Note: The plastic bottles and cups in the pictures are not included. Suggested for a group of 2 to 3 students

  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models

Specifications

Grade 5+
Class Size 1 Lab Group
Product Type Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.