Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENO1/2/3 Recombinant Rabbit Monoclonal Antibody (K01_1Q80)

Catalog No. MA565109
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA565109 50 μL
MA563650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA565109 Supplier Invitrogen™ Supplier No. MA565109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

ENO1/2/3 Recombinant Monoclonal Antibody for Western Blot

Enolase 1 is a multifunctional enzyme that, as well as its role in glycolysis, plays a part in various processes such as growth control, hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its ability to serve as a receptor and activator of plasminogen on the cell surface of several cell-types such as leukocytes and neurons. Stimulates immunoglobulin production.MBP1 binds to the myc promoter and acts as a transcriptional repressor. May be a tumor suppressor.Enolase 2 has neurotrophic and neuroprotective properties on a broad spectrum of central nervous system (CNS) neurons. Binds, in a calcium-dependent manner, to cultured neocortical neurons and promotes cell survival.Enolase 3 appears to have a function in striated muscle development and regeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENO1/2/3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q80
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ENO1
Gene Accession No. P04764, P06733, P07323, P09104, P13929, P15429, P17182, P17183, P21550
Gene Alias 0610008I15; 2-phospho-D-glycerate hydrolase; 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; AI837106; AL022784; alpha enolase; alpha enolase like 1; alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; BBE; beta-enolase; c-Myc promoter binding protein; C-myc promoter-binding protein; D6Ertd375e; Eno1; Eno-1; ENO1L1; ENO2; Eno-2; Eno3; Eno-3; Enolase 1; enolase 1, (alpha); enolase 1, alpha non-neuron; enolase 2; enolase 2 (gamma, neuronal); enolase 2, gamma neuronal; enolase 2, gamma, neuronal; enolase 3; enolase 3 (beta, muscle); enolase 3, beta muscle; enolase 3, beta, muscle; enolase-alpha; epididymis secretory protein Li 17; epididymis secretory protein Li 279; gamma-enolase; GSD13; HEL-S-17; HEL-S-279; MBP-1; MBPB1; MPB1; MPB-1; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); muscle-specific enolase; MYC promoter-binding protein 1; neural enolase; neuron specific gamma enolase; neuronal enriched enolase; neurone-specific enolase; neuron-specific enolase; NNE; Non-neural enolase; NSE; Phosphopyruvate hydratase; plasminogen-binding protein; PPH; RNEN3; Skeletal muscle enolase; tau-crystallin
Gene Symbols ENO1, ENO2, ENO3
Host Species Rabbit
Immunogen Recombinant protein of human ENO1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13806, 13807, 13808, 2023, 2026, 2027, 24333, 24334, 25438
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.