Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP2 Monoclonal Antibody (C4), Invitrogen™

Catalog No. p-7240800
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543840 20 μL
PIMA543839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543840 Supplier Invitrogen™ Supplier No. MA543840
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5' end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This gene product stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. The gene product is secreted and further processed to make the biologically active form. Several alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP2
Applications Western Blot
Classification Monoclonal
Clone A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ENPP2
Gene Accession No. Q13822
Gene Alias ATX; ATX-X; autotaxin; AutotaxinATX; autotaxin-t; ectonucleotide pyrophosphatase/phosphodiesterase 2; ectonucleotide pyrophosphatase/phosphodiesterase family member 2; E-NPP 2; Enpp2; Extracellular lysophospholipase D; FLJ26803; lysoPLD; NPP2; Npps2; PD-IALPHA; PDNP2; phosphodiesterase I/nucleotide pyrophosphatase 2; phosphodiesterase I/nucleotide pyrophosphatase 2 (autotaxin); plasma lysophospholipase D
Gene Symbols ENPP2
Host Species Mouse
Immunogen Arg637-Lys905 (Accession Q13822), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.