Learn More
Description
Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (94%), Chicken (81%), Bovine (93%). Positive Control: human cerebrospinal fluid, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ENPP2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ENPP2 |
| Gene Accession No. | Q13822 |
| Gene Alias | ATX; ATX-X; autotaxin; AutotaxinATX; autotaxin-t; ectonucleotide pyrophosphatase/phosphodiesterase 2; ectonucleotide pyrophosphatase/phosphodiesterase family member 2; E-NPP 2; Enpp2; Extracellular lysophospholipase D; FLJ26803; lysoPLD; NPP2; Npps2; PD-IALPHA; PDNP2; phosphodiesterase I/nucleotide pyrophosphatase 2; phosphodiesterase I/nucleotide pyrophosphatase 2 (autotaxin); plasma lysophospholipase D |
| Gene Symbols | ENPP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
