Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560099 Supplier Invitrogen™ Supplier No. PA560099
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKLLVFLLDG FRSDYISDEA LESLPGFKEI VSRGVKVDYL TPDFPSLSYP NYYTLMTGRH CEVHQMIGNY MWDPTTNKSF DIGVNKDSLM PLWWNGS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Choline-specific glycerophosphodiester phosphodiesterase. Hydrolyzes the classical substrate for phospholipase C, p-nitrophenyl phosphorylcholine, while it does not hydrolyze the classical nucleotide phosphodiesterase substrate, p-nitrophenyl thymidine 5'-monophosphate. Hydrolyzes lysophosphatidylcholine (LPC) to form monoacylglycerol and phosphorylcholine but not lysophosphatidic acid, showing it has a lysophospholipase C activity. Has a preference for LPC with short (12:0 and 14:0) or polyunsaturated (18:2 and 20:4) fatty acids. Also hydrolyzes glycerophosphorylcholine and sphingosylphosphorylcholine efficiently.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Enpp6
Gene Accession No. Q6UWR7
Gene Alias 4833421B01Rik; B830047L21Rik; choline-specific glycerophosphodiester phosphodiesterase; D8Ertd514e; ectonucleotide pyrophosphatase/phosphodiesterase 6; ectonucleotide pyrophosphatase/phosphodiesterase family member 6; E-NPP 6; Enpp6; E-NPP6; Glycerophosphocholine cholinephosphodiesterase; GPC-Cpde; Npp6; NPP-6; UNQ1889/PRO4334
Gene Symbols Enpp6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ENPP6. Recombinant protein control fragment (Product #RP-96724).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133121
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.