Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Enrichment Pouch
SCI_ED

Catalog No. S26747 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Includes:
Filter
No filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S26747 Filter
S26746 No filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S26747 Supplier 3M™ Supplier No. QEBPW225F
Add to Cart
Add to Cart

Ready-to-use pouch with 225 mL buffered peptone water broth. Puncture-resistant and ideal for sample preparation in environmental testing.

  • Pre-Filled, sterile
  • 225 mL buffered peptone water broth, gamma irradiated for immediate use
  • Durable Design: Heavyweight, puncture-resistant, multi-laminate polyester pouch ensures secure handling
  • Flat Bottom Stability: Stand-up pouch design provides ease during sample addition
  • Easy to Seal: Built-in wire tie keeps pouch open for filling and securely closed for transport
  • Long Shelf Life: 2 years from date of manufacture for extended storage and readiness
  • Certified Quality: ISO 9001:2008 compliant for consistent manufacturing standards Includes
  • Filter: Integrated filtration for sample processing

Specifications

Certifications/Compliance ISO 9001:2008
Color Clear
Media Type 225 mL buffered peptone water broth
Product Type Enrichment
Test Type Environmental Test
For Use With (Application) Sample preparation and enrichment procedures
Includes Filter
Packaging Type Pouch
Quantity 32/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.