Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Entactin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205738 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8994 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8994 Supplier Proteintech Group Inc Supplier No. 137661AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Although the precise composition and structure of basal laminae varies from tissue to tissue, as well as within the same tissue at different developmental stages, most mature basal laminae contain type IV collagen, laminins, heparan sulphate proteoglycans and entactin (or nidogen). Entactin is a sulfated glycoprotein that is widely distributed in basement membranes. It is tightly associated with laminin and also binds to collagen IV. Entactin probably has a role in cell-extracellular matrix interactions.

Specifications

Antigen Entactin
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NID1
Gene Accession No. P08460, P10493, P14543
Gene Alias Entactin, NID, NID 1, NID1, Nidogen 1
Gene Symbols Nid1
Host Species Rabbit
Immunogen Entactin Fusion Protein Ag4749
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18073, 25494, 4811
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.