Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Entecavir
GREENER_CHOICESDP

Catalog No. 623450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-345-0 50 mg
62-341-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-345-0 Supplier Tocris Bioscience™ Supplier No. 6234/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective hepatitis B virus inhibitor

Entecavir is a potent and selective hepatitis B virus reverse transcriptase inhibitor (EC50 = 3 nM); guanine analog. Inhibits hepatitis B virus replication in HepG2.2.15 cells. Displays selectively over HCMV, HSV-1, VZV, HIV and influenza (EC50 values are 15, 32, 30-60, >10 and >80 μM respectively). Orally bioavailable.

Specifications

CAS 142217-69-4
Quantity 50 mg
Molecular Formula C12H15N5O3
Formula Weight 277.28
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.