Learn More
3M™ Petrifilm™ Enterobacteriaceae Count Plates, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S30691 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
1000 Pack
Catalog No. Quantity
S30691 1000 Pack
1 options
Catalog No. S30691 Supplier 3M™ Petrifilm™ Supplier No. 6421
Add to Cart
Add to Cart
Cost-effective, convenient, reliable method for testing equipment, raw materials, food products and manufacturing environmental samples and bottled water testing
  • Results in 24 hours
  • Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
  • Provides accurate, efficient, consistent results for food processors and contract labs
  • Can be used with 3M™ Petrifilm™ plate reader for consistent, fast, automated reading and recording of results
  • This plate has been awarded Official Methods of Analysis (OMA) #2003.01 for selected foods and NF VALIDATION certificate: 3M™ 01/6-09/97 for All human food products, Animal feed and Industrial environmental samples by AFNOR Certification
Specifications
Detects Enterobacteriaceae
Product Type Count Plate
Test Type Food and Beverage, Environmental Test
For Use With (Application) Testing equipment, raw materials, food products and manufacturing environmental samples and bottled water for food processors and contract labs
For Use With (Equipment) 3M™ Petrifilm™ plate reader
Quantity 1000 Pack