Learn More
AdipoGen Enterocin
SDP
Supplier:  AdipoGen AGCN20116C250
Encompass_Preferred
Chemical. CAS 59678-46-5. Formula C22H20O10. MW 444.4. Isolated from Streptomyces sp. Potent antibiotic. Bacteriocin. Antibacterial. Broad spectrum activity against Gram-positive and Gram-negative bacteria. Herbicidal. Enterocin is a small molecular weight secondary metabolite and should not be confused with bacterial proteins isolated from Enterococci generically referred to as "enterocins".
Catalog No. 50-256-2655
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.