Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Enterococcus faecium derived from ATCC™ 8459™
SDP

Catalog No. 23047356 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 Pack
6 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-356 6 Pack
23-047-360 2 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-047-356 Supplier Microbiologics™ Supplier No. 01260K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simple, all-inclusive design making life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab.

  • Used as a surrogate microorganism in the pasteurization of nuts
  • Self-contained units of a single organism

Specifications

ATCC® ATCC™ 8459
Biosafety Level 1
Content And Storage 2°C to 8°C
Format KWIK-STIK
Manufacturing Origin United States
Product Type Quality Control Organism
Packaging Type Plastic Canister
Quantity 6 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.