Learn More
ATCC Enterococcus faecium (IDCC 2102) Frozen culture, BAA-3146 (ATCC)
Supplier: ATCC BAA3146/DEL
ATCC BAA-3146 (Enterococcus faecium strain IDCC 2102) is a frozen, genome-sequenced bacterial culture isolated from infant feces in South Korea for enteric and infectious disease research. It is supplied frozen for laboratory propagation and analysis.
- Genome sequenced strain.
- Isolated from infant feces in South Korea.
- Supplied frozen; recommended storage -80°C or colder.
- Grows aerobically at 37°C.
- Typical media: MRS or trypticase soy agar/broth with defibrinated sheep blood.
- Depositor: ILDONG Pharmaceutical Co., Ltd.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.