Learn More
ATCC Enterococcus faecium; TEX16
Supplier:  ATCC BAA472
Encompass
Enterococcus faecium strain TEX16 is a whole-genome sequenced bacterium that was isolated from the blood of a patient with endocarditis.
Catalog No. NC2314181
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.