Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enterovirus pan Monoclonal Antibody (L66J)
GREENER_CHOICE

Catalog No. PIMA518206 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518206 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518206 Supplier Invitrogen™ Supplier No. MA518206
  • $622.50
    $490.65 / Each of 1

    Save $131.85

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reactive with Rhinoviruses (9), Coxsackie B viruses (6), ECHO viruses (6), Coxsackie A viruses (4), EV71, and Poliovirus. Non-reactive with Influenza A, Influenza B, RSV, Adenovirus, Para 1, Para 2, Para 3, Chlamydia pneumoniae, Mycoplasma pneumoniae and Hepatitis A virus. Purity is >90%.

Enterovirus is a genus of positive-sense single-stranded RNA viruses associated with several human and mammalian diseases. Enteroviruses are named by their transmission-route through the intestine. Serologic studies have distinguished 71 human enterovirus serotypes on the basis of antibody neutralization tests. Diseases caused by enteroviruses include paralytic poliomyelitis, non-polio enteroviral infection (Enterovirus D68) and aseptic meningitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enterovirus pan
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone L66J
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias Coxsackie A; Coxsackie B; ECHO; EV71; Poliovirus; Rhinoviruses
Host Species Mouse
Immunogen Enterovirus native protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.