Learn More
Caron Products And Services Environmental Chamber, 2-Door, 50 cu. ft., Temp & RH
Supplier:  Caron Products And Services 7000502
Encompass_Preferred
50 ft3 (1,416 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.
Catalog No. NC2676671
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.