Learn More
3M™ Enviro Swab ENVSWB100, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26772 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
S26772 100 Pk.
1 options
Catalog No. S26772 Supplier 3M™ Supplier No. ENVSWB100
Add to Cart
Add to Cart
Pre-moistened swab, ready to use for collecting environmental samples on all types of food contact surfaces in the food industry
  • Ready-to-use swab reaches into corners and hard-to-reach areas for increased accuracy
  • Tamper-evident band indicates if swab has been used
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Resealable tube provides tight closure for sample transportation
  • Self-contained swab and vial for increased productivity and efficiency
Specifications
Product Type Pre-moistened Swab
For Use With (Application) Collecting environmental samples in the food industry
Quantity 100 Pk.