Learn More
International Enviroguard™ Soft Scrubs™ Disposable Pants
Catalog No. 19472728 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-728 X-Large
19-472-726 Medium
19-472-730 3X-Large
19-472-729 2X-Large
19-472-731 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-472-728 Supplier International Enviroguard™ Supplier No. FS2062BXL
This item is not returnable. View return policy
Ideal undergarments where sterile gowning is used.
Precisely cut for an easy fit
  • Soft, smooth denim blue material
  • Reduce cross-contamination
  • Elastic waist
  • Right rear hip pocket
  • Open ankles
  • Reinforced crotch
Specifications
Size X-Large
Gender Unisex
Material SMS
Color Blue
Ankle Style Open
Disposable Single-use
Sterility Non-sterile
Waist Style Elastic