Learn More
Thermco Products EnviroLog™ USB PDF Temp Logger
Catalog No. 04500043
Change view
Click to view available options
Display Type:
No Display
Catalog No. Display Type
04-500-043 No Display
1 options
Catalog No. 04-500-043 Supplier Thermco Products Supplier No. EV16
Add to Cart
Add to Cart
The EnviroLog™ USB PDF Temp Logger is ideal for monitoring temperature-sensitive goods like fresh produce pharmaceuticals and biomaterials during transportation.
  • Measures temperatures from -30°C to +70°C with an accuracy of ±0.5°C.
  • Stores up to 16,129 readings, supporting approximately 112 days at 10 min. intervals or 168 days at 15 min. intervals.
  • Features a USB 2.0 A-type plug that automatically generates PDF reports for easy data viewing in multiple languages.
  • Configurable logging intervals from 30 sec. to 18 hr., with push-button start and optional start delay from 1 min. to 72 hr.
  • Powered by a CR2032 battery providing up to 2 Years of storage life and 6 Months of operating life.
  • Integrated real-time clock with ±25 ppm accuracy ensures precise time-stamped data.
  • Housed in a durable IP65-rated case measuring 93 x 54.5 x 8.6 mm and weighing 31 g.
  • Data can be viewed in list and graph formats for efficient analysis.
  • Ideal for logistics, food, and pharmaceutical applications.
Specifications
Type Temperature Datalogger
Display Type No Display
Alarm Types Visual, LED Indicator