Learn More
Environmental Express™ SimpleDist™ System Components
Catalog No. 09735154 Shop All Environmental Express Products
Change view
Click to view available options
Quantity:
Pack of 100
Catalog No. Quantity
09-735-154 Pack of 100
1 options
Catalog No. 09-735-154 Supplier Environmental Express™ Supplier No. C6110
Add to Cart
Add to Cart
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
Specifications
Type Reagent addition tube w/funnel tip
Material Polypropylene
For Use With (Equipment) SimpleDist System
Quantity Pack of 100