Learn More
Environmental Express™ SimpleDist™ System Components
Catalog No. 09735153 Shop All Environmental Express Products
Change view
Click to view available options
Capacity (Metric):
25 mL
Catalog No. Capacity (Metric)
09-735-153 25 mL
1 options
Catalog No. 09-735-153 Supplier Environmental Express™ Supplier No. C6100
Add to Cart
Edge
Add to Cart
Ensure cyanide and ammonia collection run smoothly by replacing parts when appropriate
  • Replacement manifolds and washers
  • Tubing and adapters can be replaced after wear and tear
  • Collection traps for safe pouring of the final solution
Specifications
Includes Built-in vent tube
Type Collection Trap
Material Polystyrene
Capacity (Metric) 25 mL
For Use With (Equipment) SimpleDist Systems